COM448631904 Combi-Blocks, Inc.

3-Nitrobenzenesulfonyl fluoride

  • MDL number MFCD00024845
  • Cas Number : 349-78-0
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-3027

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-3027
Availability details for
Quantity
  • Availability
  • Add to Cart