COM448631906 Combi-Blocks, Inc.

4-Chloro-3-nitrobenzenesulfonyl fluoride

  • MDL number MFCD00024857
  • Cas Number : 349-04-2
  • Molecular Weight : 239.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-3080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-3080
Availability details for
Quantity
  • Availability
  • Add to Cart