COM448631913 Combi-Blocks, Inc.

4-(Benzyloxy)-2-bromobenzamide

  • MDL number MFCD28369589
  • Cas Number : 1820665-08-4
  • Molecular Weight : 306.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4001

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4001
Availability details for
Quantity
  • Availability
  • Add to Cart