COM448631917 Combi-Blocks, Inc.

4-(4-Bromophenyl)-1-methyl-3,6-dihydro-2H-pyridine

  • MDL number MFCD27946380
  • Cas Number : 180912-08-7
  • Molecular Weight : 252.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4005

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4005
Availability details for
Quantity
  • Availability
  • Add to Cart