COM448631931 Combi-Blocks, Inc.

1-(1-Bromo-3-chloropropyl)-4-chlorobenzene

  • MDL number MFCD25966870
  • Cas Number : 745817-23-6
  • Molecular Weight : 267.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4029

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4029
Availability details for
Quantity
  • Availability
  • Add to Cart