COM448631951 Combi-Blocks, Inc.

4-Thiophen-2-ylphenol

  • MDL number MFCD06797407
  • Cas Number : 29886-65-5
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-8018

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-8018
Availability details for
Quantity
  • Availability
  • Add to Cart