COM448631961 Combi-Blocks, Inc.

4-Bromo-2-(BOC-amino)methylthiophene

  • MDL number MFCD10700150
  • Cas Number : 479090-39-6
  • Molecular Weight : 292.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-8038
Availability details for
Quantity
  • Availability
  • Add to Cart