COM448632017 Combi-Blocks, Inc.

N-(Ethanesulfonyl)piperidine-4-boronic acid pinacol ester

  • MDL number MFCD28166395
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1000

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1000
Availability details for
Quantity
  • Availability
  • Add to Cart