COM448632026 Combi-Blocks, Inc.

2-(4-Bromophenylthio)-5-nitrophenylboronic acid pinacol ester

  • MDL number MFCD28166401
  • Molecular Weight : 436.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1029

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1029
Availability details for
Quantity
  • Availability
  • Add to Cart