COM448632027 Combi-Blocks, Inc.

4-Methoxy-2-(methoxycarbonyl)pyridine-5-boronic acid pinacol ester

  • MDL number MFCD18727245
  • Molecular Weight : 293.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1030

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1030
Availability details for
Quantity
  • Availability
  • Add to Cart