COM448632028 Combi-Blocks, Inc.

3-(BOC-Amino)-2-methoxycarbonylpyridine-5-boronic acid pinacol ester

  • MDL number MFCD28369591
  • Cas Number : 2377606-60-3
  • Molecular Weight : 378.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1031
Availability details for
Quantity
  • Availability
  • Add to Cart