COM448632034 Combi-Blocks, Inc.

3-Methylcyclohex-1-ene-2-boronic acid pinacol ester

  • MDL number MFCD22494818
  • Cas Number : 448211-44-7
  • Molecular Weight : 222.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 97
Storage temp. Freezer

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1047
Availability details for
Quantity
  • Availability
  • Add to Cart