COM448632040 Combi-Blocks, Inc.

tert-Butyl 4-chloro-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)indazole-1-carboxylate

  • MDL number MFCD28397292
  • Cas Number : 2377611-95-3
  • Molecular Weight : 378.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1070

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1070
Availability details for
Quantity
  • Availability
  • Add to Cart