COM448632041 Combi-Blocks, Inc.

3-methylbenzylboronic acid pinacol ester

  • MDL number MFCD23102598
  • Cas Number : 365564-12-1
  • Molecular Weight : 232.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0034

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0034
Availability details for
Quantity
  • Availability
  • Add to Cart