COM448632049 Combi-Blocks, Inc.

5-Methoxypyridine-2-boronic acid pinacol ester

  • MDL number MFCD08752652
  • Cas Number : 1309601-77-1
  • Molecular Weight : 235.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0141
Availability details for
Quantity
  • Availability
  • Add to Cart