COM448632059 Combi-Blocks, Inc.

3-(1,1-Dioxido-2-isothiazolidinyl)phenylboronic acid pinacol ester

  • MDL number MFCD28024844
  • Cas Number : 1416367-18-4
  • Molecular Weight : 323.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0543

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0543
Availability details for
Quantity
  • Availability
  • Add to Cart