COM448632065 Combi-Blocks, Inc.

2,2-Dimethyl-1-[4-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazin-1-yl]propan-1-one

  • MDL number MFCD24039610
  • Molecular Weight : 372.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0740
Availability details for
Quantity
  • Availability
  • Add to Cart