COM448632074 Combi-Blocks, Inc.

4,4,5,5-Tetramethyl-2-[2-(3-nitrophenoxy)ethyl]-1,3,2-dioxaborolane

  • MDL number MFCD27935917
  • Molecular Weight : 293.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0749

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0749
Availability details for
Quantity
  • Availability
  • Add to Cart