COM448632084 Combi-Blocks, Inc.

Acetamide, n-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)phenyl]-

  • MDL number MFCD22573204
  • Cas Number : 1374606-49-1
  • Molecular Weight : 329.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0759

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0759
Availability details for
Quantity
  • Availability
  • Add to Cart