COM448632085 Combi-Blocks, Inc.

5-Chloro-4-methyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • MDL number MFCD24039501
  • Cas Number : 2377608-30-3
  • Molecular Weight : 267.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0760

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0760
Availability details for
Quantity
  • Availability
  • Add to Cart