COM448632087 Combi-Blocks, Inc.

4-Chloro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)aniline

  • MDL number MFCD24039570
  • Cas Number : 1995072-92-8
  • Molecular Weight : 321.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0762

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0762
Availability details for
Quantity
  • Availability
  • Add to Cart