COM448632107 Combi-Blocks, Inc.

4-Methoxy-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • MDL number MFCD24039683
  • Cas Number : 2088729-76-2
  • Molecular Weight : 249.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0788

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0788
Availability details for
Quantity
  • Availability
  • Add to Cart