COM448632108 Combi-Blocks, Inc.

N-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethoxy)phenyl]acetamide

  • MDL number MFCD24039372
  • Molecular Weight : 345.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0789

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0789
Availability details for
Quantity
  • Availability
  • Add to Cart