COM448632110 Combi-Blocks, Inc.

1-[2-Nitro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-one

  • MDL number MFCD27935937
  • Molecular Weight : 305.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0791

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0791
Availability details for
Quantity
  • Availability
  • Add to Cart