COM448632113 Combi-Blocks, Inc.

4,4,5,5-Tetramethyl-2-(5-nitro-2-propoxyphenyl)-1,3,2-dioxaborolane

  • MDL number MFCD27935939
  • Molecular Weight : 307.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0794

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0794
Availability details for
Quantity
  • Availability
  • Add to Cart