COM448632130 Combi-Blocks, Inc.

2-Nitro-5-(pentyloxy)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • MDL number MFCD27935950
  • Molecular Weight : 350.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0811

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0811
Availability details for
Quantity
  • Availability
  • Add to Cart