COM448632159 Combi-Blocks, Inc.

1-N-Ethyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine

  • MDL number MFCD27935970
  • Molecular Weight : 262.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0840

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0840
Availability details for
Quantity
  • Availability
  • Add to Cart