COM448632167 Combi-Blocks, Inc.

2,3,5,6-Tetramethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • MDL number MFCD24039526
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0848

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0848
Availability details for
Quantity
  • Availability
  • Add to Cart