COM448632172 Combi-Blocks, Inc.

tert-Butyl 4-[4-amino-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine-1-carboxylate

  • MDL number MFCD24039356
  • Molecular Weight : 403.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0853

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0853
Availability details for
Quantity
  • Availability
  • Add to Cart