COM448632192 Combi-Blocks, Inc.

1-N-Cyclopentyl-5-fluoro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine

  • MDL number MFCD24039666
  • Molecular Weight : 320.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0873

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0873
Availability details for
Quantity
  • Availability
  • Add to Cart