COM448632199 Combi-Blocks, Inc.

2-(2-Ethoxy-5-fluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD27935998
  • Molecular Weight : 311.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0880

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0880
Availability details for
Quantity
  • Availability
  • Add to Cart