COM448632202 Combi-Blocks, Inc.

N-Cyclohexyl-2-nitro-5-propoxy-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • MDL number MFCD27936001
  • Molecular Weight : 404.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0883

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0883
Availability details for
Quantity
  • Availability
  • Add to Cart