COM448632213 Combi-Blocks, Inc.

2-Ethoxy-3-methyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • MDL number MFCD24039673
  • Molecular Weight : 277.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0894

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0894
Availability details for
Quantity
  • Availability
  • Add to Cart