COM448632218 Combi-Blocks, Inc.

2-[5-(Benzyloxy)-2-chloro-4-nitrophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD27936015
  • Molecular Weight : 389.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0899
Availability details for
Quantity
  • Availability
  • Add to Cart