COM448632223 Combi-Blocks, Inc.

2-(5-Fluoro-2-isopropoxy-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD27936020
  • Molecular Weight : 325.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0904

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0904
Availability details for
Quantity
  • Availability
  • Add to Cart