COM448632236 Combi-Blocks, Inc.

4-Chloro-2-nitro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

  • MDL number MFCD27936033
  • Molecular Weight : 327.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0918
Availability details for
Quantity
  • Availability
  • Add to Cart