COM448632237 Combi-Blocks, Inc.

5-Chloro-2-propoxy-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)aniline hydrochloride

  • MDL number MFCD27936034
  • Molecular Weight : 348.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0919
Availability details for
Quantity
  • Availability
  • Add to Cart