COM448632251 Combi-Blocks, Inc.

1,3,2-Dioxaborolane, 4,4,5,5-tetramethyl-2-[2-(4-nitrophenoxy)phenyl]-

  • MDL number MFCD22415461
  • Cas Number : 870221-30-0
  • Molecular Weight : 341.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0933

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0933
Availability details for
Quantity
  • Availability
  • Add to Cart