COM448632260 Combi-Blocks, Inc.

3-Amino-4-fluoro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

  • MDL number MFCD27936053
  • Molecular Weight : 281.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0942
Availability details for
Quantity
  • Availability
  • Add to Cart