COM448632269 Combi-Blocks, Inc.

5-Amino-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-ol

  • MDL number MFCD26519964
  • Cas Number : 1787251-98-2
  • Molecular Weight : 236.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0951

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0951
Availability details for
Quantity
  • Availability
  • Add to Cart