COM448632275 Combi-Blocks, Inc.

4-[2-(Tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]aniline hydrochloride

  • MDL number MFCD27936065
  • Molecular Weight : 283.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0957

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0957
Availability details for
Quantity
  • Availability
  • Add to Cart