COM448632280 Combi-Blocks, Inc.

2-hydroxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile

  • MDL number MFCD18724096
  • Cas Number : 2256755-07-2
  • Molecular Weight : 246.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0962

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0962
Availability details for
Quantity
  • Availability
  • Add to Cart