COM448632295 Combi-Blocks, Inc.

2-Nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzeneacetic acid, methyl ester

  • MDL number MFCD19689542
  • Cas Number : 1352413-08-1
  • Molecular Weight : 321.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0978

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0978
Availability details for
Quantity
  • Availability
  • Add to Cart