COM448632296 Combi-Blocks, Inc.

Benzoic acid, 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 1,1-dimethylethyl ester

  • MDL number MFCD22494336
  • Cas Number : 1351501-00-2
  • Molecular Weight : 322.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0979

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0979
Availability details for
Quantity
  • Availability
  • Add to Cart