COM448632297 Combi-Blocks, Inc.

tert-Butyl 4-fluoro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • MDL number MFCD24040014
  • Cas Number : 2377607-40-2
  • Molecular Weight : 322.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0980
Availability details for
Quantity
  • Availability
  • Add to Cart