COM448632314 Combi-Blocks, Inc.

5-Fluoro-n-(2-methylpropyl)-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD24039561
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0997
Availability details for
Quantity
  • Availability
  • Add to Cart