COM448632318 Combi-Blocks, Inc.

Methyl 4-chloro-2-hydroxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • MDL number MFCD24039379
  • Cas Number : 1820813-94-2
  • Molecular Weight : 312.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1001

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1001
Availability details for
Quantity
  • Availability
  • Add to Cart