COM448632327 Combi-Blocks, Inc.

1-[[3-Methyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbonyl]pyrrolidine

  • MDL number MFCD24039629
  • Cas Number : 1386860-56-5
  • Molecular Weight : 315.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1010

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1010
Availability details for
Quantity
  • Availability
  • Add to Cart