COM448632336 Combi-Blocks, Inc.

Methyl 4-nitro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • MDL number MFCD27936077
  • Cas Number : 2377608-56-3
  • Molecular Weight : 307.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1019

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1019
Availability details for
Quantity
  • Availability
  • Add to Cart