COM448632337 Combi-Blocks, Inc.

6-(Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carbohydrazide

  • MDL number MFCD24039896
  • Molecular Weight : 312.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1020

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1020
Availability details for
Quantity
  • Availability
  • Add to Cart